Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Phenylthiophene-2-carboxylic acid, 95+%, Thermo Scientific™

Catalog No. CC10501DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC10501DA 1 g Amber Glass Bottle
CC10501CB 250 mg Amber Glass Bottle
CC10501DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC10501DA Supplier Thermo Scientific Chemicals Supplier No. CC10501DA
Add to Cart

Chemical Identifiers

CAS 19163-24-7
Molecular Formula C11H8O2S
Molecular Weight (g/mol) 204.243
InChI Key QGMFBCDNJUZQBZ-UHFFFAOYSA-N
Synonym 5-phenyl-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-phenyl, 5-phenyl-2-thiophene carboxylic acid, pubchem19533, 5-phenyl-2-thenoic acid, 2-carboxy-5-phenylthiophene, 5-phenyl 2-thiophenecarboxylic acid, 5-phenyl-thiophene-2-carboxylic acid
PubChem CID 736494
IUPAC Name 5-phenylthiophene-2-carboxylic acid
SMILES C1=CC=C(C=C1)C2=CC=C(S2)C(=O)O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 204.25
Percent Purity 95%
Chemical Name or Material 5-Phenylthiophene-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.