Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroquinine, 95%

Catalog No. AC461650050
Click to view available options
Quantity:
5 g

CAS: 522-66-7 | C20H26N2O2 | 326.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 522-66-7
Molecular Formula C20H26N2O2
Molecular Weight (g/mol) 326.44
MDL Number MFCD00151107
InChI Key LJOQGZACKSYWCH-ADMHDHIONA-N
Synonym dihydroquinine, hydroquinine, 1r-2s,4s,5r-5-ethylquinuclidin-2-yl 6-methoxyquinolin-4-yl methanol, unii-31j3q51t6l, hydrochinin, quinine, 10,11-dihydro, --dihydroquinine, --dihydroquinine, analytical standard, 8a,9r-10,11-dihydro-6'-methoxycinchonan-9-ol
PubChem CID 121515
IUPAC Name (R)-[(2S,4S,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol
SMILES CCC1CN2CCC1CC2[C@H](O)C1=CC=NC2=CC=C(OC)C=C12

Specifications

Melting Point 168.0°C
Boiling Point 468.4°C
Flash Point 225°C
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 326.43
Percent Purity 95%
Chemical Name or Material Hydroquinine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
EINECSNumber 208-334-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.