Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Pyrimidinecarboxaldehyde, 95%, Thermo Scientific™

Catalog No. AC454300010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454300010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC454300010 Supplier Thermo Scientific Chemicals Supplier No. 454300010

Chemical Identifiers

CAS 10070-92-5
Molecular Formula C5H4N2O
Molecular Weight (g/mol) 108.10
MDL Number MFCD03426065
InChI Key FREJAOSUHFGDBW-UHFFFAOYSA-N
Synonym pyrimidine-5-carboxaldehyde, 5-formylpyrimidine, pyrimidine-5-carboxyaldehyde, 5-pyrimidinecarboxaldehyde, 5-pyrimidinecarbaldehyde, 5-pyrimidinaldehyde, 5-formyl pyrimidine, pubchem5298, pyrimidine-5-aldehyde, pyrimidine 5-carboxaldehyde
PubChem CID 2761034
IUPAC Name pyrimidine-5-carbaldehyde
SMILES O=CC1=CN=CN=C1

Specifications

Melting Point 39.0°C to 43.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 108.1
Percent Purity 95%
Chemical Name or Material 5-Pyrimidinecarboxaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.