Learn More
5-Sulfoisophthalic acid monosodium salt, 95%
Description
It can be used as the third monomer in the Polyester procession of esterification, which can improve the ability of product's dying. And it can be used as the catalyst in medicine, agricultural chemical, polyester. Used as a monomer for waterborne polyesters and nylon fibers, in adhesion aid for magnetic tapes, and amine free polyurethane dispersions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It can be used as the third monomer in the Polyester procession of esterification, which can improve the ability of product′s dying. And it can be used as the catalyst in medicine, agricultural chemical, polyester. Used as a monomer for waterborne polyesters and nylon fibers, in adhesion aid for magnetic tapes, and amine free polyurethane dispersions.
Solubility
Soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 100 g |
| Beilstein | 4170770 |
| Solubility Information | Soluble in water. |
| Formula Weight | 268.18 |
| Percent Purity | 95% |
| Chemical Name or Material | 5-Sulfoisophthalic acid monosodium salt |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 228-845-2 |
| RTECSNumber | DB6030000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.