Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-SulfoSalicylic Acid Dihydrate, ACS Reagent, Reagents

Catalog No. 01337648
Click to view available options
Quantity:
125 g
500 g
3 kg
10 kg

5-SulfoSalicylic Acid Dihydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 5965-83-3
Molecular Formula C9H14O6S
Molecular Weight (g/mol) 250.27
InChI Key NFYHZVWMQHQKRU-UHFFFAOYSA-N
Synonym 2-Hydroxy-5-sulfobenzoic Acid dihydrate, Sulfosalicylic Acid dihydrate
IUPAC Name 2-hydroxy-5-sulfobenzoic acid; bis(methane)
SMILES C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O

Specifications

Color White
Packaging Poly Bottle
Quantity 500 g
Specific Gravity 0.8
Grade ACS
Physical Form Solid
Chemical Name or Material 5-SulfoSalicylic Acid Dihydrate, ACS Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.