Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-Sulfosalicylic Acid Dihydrate (Crystalline/Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
3 kg
Packaging:
Glass Bottle
Poly Bottle
Specifications
Specifications
Alpha Vector | Sulfosalicylic Acid Dihydrate |
Melting Point | 106°C |
Color | White |
pH | 0.5 |
Ignition Residue | 0.1% max. |
Assay Percent Range | 99.0 to 101.0% |
Packaging | Poly Bottle |
Linear Formula | 1-(OH)-2-(COOH)-4-SO3H-C6H3·2H2O |
Quantity | 3 kg |
UN Number | 2585 |
Show More |
Safety and Handling

DOTInformation | DOT Class 8, : Corrosive |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction