Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(thien-2-yl)thiophene-2-carbonitrile, Thermo Scientific™

Catalog No. p-4391483
Click to view available options
Quantity:
250 mg
5 g

Chemical Identifiers

CAS 16278-99-2
Molecular Formula C9H5NS2
Molecular Weight (g/mol) 191.266
InChI Key ANARYBGZNUMARH-UHFFFAOYSA-N
Synonym 5-thien-2-yl thiophene-2-carbonitrile, 2,2'-bithiophene-5-carbonitrile, 5-thiophen-2-yl thiophene-2-carbonitrile, maybridge3_006613, 5-cyano-2,2'-bithiophene, 5-2-thienyl thiophene-2-carbonitrile
PubChem CID 2739839
IUPAC Name 5-thiophen-2-ylthiophene-2-carbonitrile
SMILES C1=CSC(=C1)C2=CC=C(S2)C#N

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 191.28
Percent Purity 97%
Chemical Name or Material 5-(Thien-2-yl)thiophene-2-carbonitrile
Hazard Category GHS Signal Word: Warning
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.