Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
50% Acetic Acid Aqueous Solution, Cenmed™

Click to view available options
Quantity:
4 x 60 mL
Description
- Packaged in 60 mL plastic vials for enhanced usability, providing convenience in handling and dispensing the solution with precision and minimizing the risk of errors
- With a precisely balanced 50% concentration, this solution delivers optimal performance, guaranteeing accuracy in your chemical experiments
- Differentiate your lab work with a reliable and versatile reagent that combines the purity of acetic acid with the convenience of an easily measured 60 mL portion
- Ensure quality in your chemical processes with a solution designed to meet the rigorous standards of precise laboratory work
- Optimize your laboratory performance with a solution that prioritizes accuracy, reliability, and convenience for controlled chemical applications
Specifications
Specifications
| Physical Form | Aqueous Solution, Liquid |
| CAS | 64-19-7 |
| Packaging | Plastic Vial |
| Color | Clear |
| Quantity | 4 x 60 mL |
| Grade | Reagent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction