Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Troemner™ 50 g-5 mg Analytical Precision UltraClass Weight Set
Shop All Troemner Products
Click to view available options
Certifications/Compliance:
NVLAP Accredited Certificate
No Certificate
Traceable Certificate
Description
- Used in extremely stable environments
- 1 to 500 mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
- Weights 1 g and above are of two-piece construction
- Weight sets 1 mg-500 g are provided in a high-quality polycarbonate set case

Specifications
Specifications
Certifications/Compliance | Traceable Certificate |
Classification | UltraClass |
Material | Troemner Alloy 8, Stainless Steel |
Case Material | Polycarbonate |
Density | Weights 1 g and above are 8.03 g/cm3 at 20°C. Weights below 1 g are 7.95 g/cm3 at 20°C. |
Mass | 50 g to 5 mg |
Length (Metric) | 20.32 cm |
Height (Metric) Case | 8.89 cm |
For Use With (Application) | Reference standard calibration |
For Use With (Equipment) | Calibration of high-precision analytical balances with readability as low as 0.01 mg |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction