Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ 50% Hydroxylamine Buffer, 5 mL PROMO
Click to view available options
:
Thermo Scientific™ 50% Hydroxylamine Buffer
Description
Triethylamonium bicarbonate (TEAB) is a 10X dissolution buffer for isobaric mass tag labeling experiments. 50% hydroxylamine is a 10X quenching reagent for amine-reactive labeling for TMT experiments.
Specifications
Specifications
| Product Type | Hydroxylamine |
| Product Line | TMT™ |
| Content And Storage | Store protected from direct sunlight in a dry, cool, and well-ventilated area. |
| Starting Material | Cell Lysate |
| Workflow Step | Protein Labeling |
| Quantity | 5 mL |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction