Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSP60 Polyclonal Antibody
Chicken Polyclonal Antibody
Supplier: Invitrogen™ PA5143537
Description
HSP60 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a member of the chaperonin family. The encoded mitochondrial protein may function as a signaling molecule in the innate immune system. This protein is essential for the folding and assembly of newly imported proteins in the mitochondria. This gene is adjacent to a related family member and the region between the 2 genes functions as a bidirectional promoter. Several pseudogenes have been associated with this gene. Two transcript variants encoding the same protein have been identified for this gene. Mutations associated with this gene cause autosomal recessive spastic paraplegia 13.
Specifications
| HSP60 | |
| Polyclonal | |
| Unconjugated | |
| HSPD1 | |
| 60 kDa chaperonin; 60 kDa heat shock protein; 60 kDa heat shock protein, mitochondrial; 60kDa; Chaperonin 60; CPN60; GROEL; heat shock 60kD protein 1 (chaperonin); heat shock 60kDa protein 1; heat shock 60kDa protein 1 (chaperonin); Heat shock protein; heat shock protein 1 (chaperonin); heat shock protein 60; heat shock protein 60 (liver); heat shock protein 65; heat shock protein family D (Hsp60) member 1; heat shock protein, 60 kDa; HLD4; HSP; Hsp60; HSP-60; Hsp60s1; HSP65; HSP-65; Hspd1; Hspd1-30p; HuCHA60; hypothetical protein; I79_011398; mitochondrial matrix protein P1; P1 protein; P60 lymphocyte protein; RCJMB04_7g5; short heat shock protein 60 Hsp60s1; SPG13; Unknown (protein for IMAGE:7943409) | |
| Chicken | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgY |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Conc. Not Determined | |
| PBS with 0.02% sodium azide | |
| P10809, P63038, P63039 | |
| HSPD1 | |
| Recombinant full length human HSP60 expressed in and purified from E. coli. | |
| RUO | |
| 15510, 3329, 63868 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction