Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 7 15,000 D MWCO Standard RC Pre-Treated Dialysis Kits
Shop All Spectrum Medical Industries Products

Click to view available options
Diameter (Metric):
7.5 mm
15 mm
Length (Metric):
0.45 mL/cm vol./length
1.8 mL/cm vol./length
Description
Includes
- 1meter of pre-treated tubing, a pair of closures and 5 opening picks
- Hydrophilic, symmetric porositiy
- Equilibrated in water
- Packaged wet in preservative solution of 0.05% sodium azide
- For small experiments or qualifying a membrane for a particular application
Specifications
Specifications
MWCO | 15 kDa |
Diameter (Metric) | 7.5 mm |
Length (Metric) | 0.45 mL/cm vol./length |
Quantity | 1 m Tubing/Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction