Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Labs Spectra/Por™ 7 25,000 D MWCO Standard RC Pre-Treated Dialysis Kits
SDP

Catalog No. p-5013613 Shop All Spectrum Medical Industries Products
Click to view available options
Diameter (Metric):
18 mm
22 mm
Length (Metric):
2.5 mL/cm vol./length
3.7 mL/cm vol./length
This item is not returnable. View return policy

Catalog No. 08-700-221

Supplier: Spectrum™ 132127T

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

  • 1meter of pre-treated tubing, a pair of closures and 5 opening picks

With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds

  • Hydrophilic, symmetric porositiy
  • Equilibrated in water
  • Packaged wet in preservative solution of 0.05% sodium azide
  • For small experiments or qualifying a membrane for a particular application

Specifications

MWCO 25 kDa
Diameter (Metric) 22 mm
Length (Metric) 3.7 mL/cm vol./length
Quantity 1 m Tubing/Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.