Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spectrum™ Labs 8 - 10 kD Biotech CE Dialysis Kits
Shop All Spectrum Medical Industries Products

Click to view available options
Diameter (Metric):
6.4 mm
15 mm
20 mm
Length (Metric):
0.32 mL/cm vol./length
1.8 mL/cm vol./length
3.1 mL/cm vol./length
Description
Includes
- 1m Biotech CE Tubing in 0.05% sodium azide
- Two universal tubing closures
- Five opening picks
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
- Hydrophilic, symmetric porositiy
- For small tubing demand applications or evaluating a specific dialysis membrane
- Includes 1m tubing with pair of tubing closures and 5 opening picks
Specifications
Specifications
MWCO | 8 to 10 kDa |
Diameter (Metric) | 20 mm |
Length (Metric) | 3.1 mL/cm vol./length |
Quantity | 1 m Tubing/Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction