Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Labs Spectra/Por™ 6 10 kD MWCO Standard RC Pre-wetted Dialysis Kits
SDP

Catalog No. p-5048677 Shop All Spectrum Medical Industries Products
Click to view available options
Diameter (Metric):
7.5 mm
15 mm
20.4 mm
29 mm
Length (Metric):
0.45 mL/cm vol./length
1.8 mL/cm vol./length
3.3 mL/cm vol./length
6.4 mL/cm vol./length
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

  • 1 meter of Spectra/Por 6 Dialysis Tubing
  • 1 Standard Spectra/Por Closure (for buoyancy)
  • 1 Weighted Spectra/Por Closure (for orientation)
  • 5 Tubing Opening Picks

With Pre-wetted ready-to-use tubing, not requiring soaking to remove glycerol

  • Clear, flexible, study standard grade Regenerated Cellulose (RC) dialysis membrane
  • Membrane packaged wet in preservative solution of 0.05% sodium azide
  • Kits available in 1m tubing lengths ideal for membrane evaluation and qualification
  • Include a pair of closures and convenient tubing opening picks

Specifications

MWCO 10 kDa
Diameter (Metric) 20.4 mm
Length (Metric) 3.3 mL/cm vol./length
Quantity 1 m Tubing/Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.