Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PP2B, Human, Recombinant Protein, E. coli

Catalog No. 5395685000U Shop All MilliporeSigma Products
Click to view available options
Quantity:
5000 U
Recombinant, human calcineurin Aα (catalytic subunit) and calcineurin B (regulatory subunit) co-expressed with yeast myristoyl-CoA:protein N-myristoyltransferase. A Ca2+/CaM-dependent protein phosphatase. The resulting highly active calcineurin is N-myristoylated on the CaN B-subunit, as is the native protein. M.W. 60,000 (A subunit) and 15,000 (B subunit).
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 100mM NaCl, 50mM Tris, 6mM MgCl2, 5mM DTT, 0.5mM CaCl2, 0.025% NP-40.
Purification Method Two distinct bands by SDS-PAGE
Quantity 5000 U
Source E. coli
Immunogen PP2B
Storage Requirements Following initial thaw, aliquot and freeze (−70°C). Please refer to vial label for lot-specific reconstitution volume.
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.