Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
53BP1 Antibody (1285A), Alexa Fluor 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals MAB18771AF750
Description
53BP1 Monoclonal specifically detects 53BP1 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
53BP1 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
TP53BP1 | |
Recombinant monoclonal 53BP1 Antibody (1285A) was made to amino acids 350 - 400 of human 53BP1 (NP_005648.1). | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
1285A | |
Western Blot, Immunocytochemistry | |
53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1 | |
Rabbit | |
Protein A or G purified | |
Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Protein Phosphatase, Tumor Suppressors, Unfolded Protein Response (UPR) | |
7158.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction