Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

53BP1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB100304 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100304 0.1 mL
NB100304SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100304 Supplier R&D Systems Supplier No. NB100304
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

53BP1 Polyclonal specifically detects 53BP1 in Avian, Bat, Bovine, Canine, Drosophila melanogaster, Fish, Goat, Human, Mouse, Naked mole-rat, Pig, Primate, Rabbit, Rat, Reptile samples. It is validated for Chemotaxis, ChIP Assay, FISH, Flow Cytometry, Flow Cytometry (Intracellular Staining), Gene Knock-Out, IF-FISH, IF/ICC, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization, In-situ Hybridization, KnockDown, Proximity Ligation Assay, Western Blot.

Specifications

Antigen 53BP1
Applications Chemotaxis, ChIP Assay, FISH, Flow Cytometry, Flow Cytometry (Intracellular Staining), Gene Knock-Out, FISH, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), In Situ Hybridization (ISH), KnockDown, Proximity Ligation Assay, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:25000, Chromatin Immunoprecipitation reported in scientific literature (PMID 24591601), Flow Cytometry 1.5 ug/ml, Immunohistochemistry reported in scientific literature (PMID 24987917), Immunocytochemistry/Immunofluorescence 1:1000-1:5000, Immunoprecipitation reported in scientific literature (PMID 25645366), Immunohistochemistry-Paraffin 1:1000-1:5000. Use reported in scientific literature (PMID 27653664), Immunohistochemistry-Frozen reported by customer review, Immunoblotting reported in multiple pieces of scientific literature, In-situ Hybridization reported in scientific literature (PMID 34988401; 33122290), Flow (Intracellular), Chromatin Immunoprecipitation (ChIP), Knockout Validated reported in scientific literature (PMID 26601238), KnockDown Validated
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q12888
Gene Alias 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1
Gene Symbols TP53BP1
Host Species Rabbit
Immunogen The epitope recognized by 53BP1 Antibody maps to a region between residues 350 and 400 of human 53BP1 [NP_005648.1].
Molecular Weight of Antigen 214 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7158
Test Specificity Human, mouse, rat and goat. Human has been tested in WB, IHC-P and ICC/IF, mouse and goat have only been tested in ICC/IF. Primate reactivity reported in scientific literature (PMID: 18389475).Fish reactivity reported in scientific literature (PMID: 25516420).
Target Species Avian, Mouse, Bat, Bovine, Canine, Canine, Drosophila melanogaster, Goat, Fish, Goat, Human, Mouse, Naked mole-rat, Pig, Primate, Rabbit, Rat, Reptile
Content And Storage Store at -20 degreesC.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.