Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
53BP1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100305MFV450
Description
53BP1 Polyclonal antibody specifically detects 53BP1 in Human, Mouse, Rat, Bat, Bovine, Canine, Naked mole-rat samples. It is validated for Flow CytometrySpecifications
53BP1 | |
Polyclonal | |
Flow Cytometry | |
53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1 | |
The epitope recognized by 53BP1 Antibody maps to a region between residues 1925 and the C-terminus (residue 1972) of human 53BP1 (NP_005648.1). | |
0.1 mL | |
Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Tumor Suppressors | |
7158 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bat, Bovine, Canine, Naked mole-rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction