Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
53BP1 Antibody, PE/Atto594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100304PEATT594
Description
53BP1 Polyclonal specifically detects 53BP1 in Human, Mouse, Rat, Bat, Bovine, Canine, Fish, Goat, Primate samples. It is validated for Flow Cytometry, Flow (Intracellular), Knockout Validated, Knockdown Validated.Specifications
53BP1 | |
Polyclonal | |
Flow Cytometry, Flow (Intracellular), Knockout Validated, Knockdown Validated | |
53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1 | |
Rabbit | |
Affinity Purified | |
Breast Cancer, Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Phospho Specific, Protein Phosphatase, Tumor Suppressors, Unfolded Protein Response (UPR) | |
7158.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry, Immunoassay, KnockDown | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
TP53BP1 | |
Partial synthetic peptide made to an internal portion of human 53BP1 (between amino acids 350-400) [NP_005648.1]. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Bat, Bovine, Canine, Fish, Goat, Primate | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction