Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

53BP1, p Ser25 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001803 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001803 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001803 Supplier Novus Biologicals Supplier No. NB1001803
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 13 publications

53BP1 Polyclonal specifically detects 53BP1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen 53BP1 (p Ser25)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Flow Cytometry 25 ug per 1 million cells in a 150 mcl reaction, Immunohistochemistry 1:1000 - 1:5000, Immunocytochemistry/Immunofluorescence 1:500 - 1:2500, Immunoprecipitation 1-4 ug/mg of lysate, Immunohistochemistry-Paraffin 1:1000 - 1:5000
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q12888
Gene Alias 53BP1, p202, p53-binding protein 1, p53bp1, TDRD30, TP53-Binding Protein 1, tumor protein 53-binding protein, 1, tumor protein p53 binding protein 1, tumor protein p53-binding protein, 1, tumor suppressor p53-binding protein 1
Gene Symbols TP53BP1
Host Species Rabbit
Immunogen 53BP1 [p Ser25] Antibody was made to a phosphorylated synthetic peptide, which represented a portion of human p53 Binding Protein 1 (GeneID 7158) around serine 25 according to the numbering given in entry NP_005648.1.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 7158
Test Specificity Based on 100% sequence identity, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Nine-banded armadillo, White-tufted-ear marmoset, Crab-eating macaque, Olive baboon and Greater horseshoe bat.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.