Learn More
Invitrogen™ 53BP1 Polyclonal Antibody, DyLight™ 550
GREENER_CHOICE
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA522794
Catalog No. PIPA522794
Only null left
Add to Cart
Description
Description
This antibody reacts with Human samples in Western blot and ICC applications, and with Mouse in ICC applications. Predicted cross-reactivity based on sequence identity: Chimpanzee/Orangutan (99%), Gibbon/Gorilla (98%), Marmoset (95%).
53BP1 (P53-binding protein 1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, 53BP1 is phosphorylated and localizes to the presumptive sites of damage, specifically, double-strand breaks. 53BP1 may have a role in checkpoint signaling during mitosis, enhances TP53-mediated transcriptional activation, and participates in DNA repair by maintaining genomic stability.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
53BP1
Polyclonal
DyLight 550
TP53BP1
53BP1; FLJ41424; m53BP1; MGC138366; murine p53-binding protein; p202; p53-binding protein 1; p53BP1; TDRD30; TP53; TP53-binding protein 1; Tp53bp1; transformation related protein 53 binding protein 1; transformation related protein 53 binding-protein 1; Trp53bp1; tumor protein 53-binding protein, 1; tumor protein p53 binding protein 1; tumor protein p53-binding protein, 1; tumor suppressor p53-binding protein 1
Rabbit
Antigen affinity chromatography
RUO
27223, 7158
4°C, store in dark
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), Western Blot, Proximity Ligation Assay, Immunocytochemistry
0.61 mg/mL
50mM sodium borate with 0.05% sodium azide
P70399, Q12888
TP53BP1, Trp53bp1
Synthetic peptide corresponding to a portion of human 53BP1 encoded in exon 11, 12 and 19.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.