Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 5730409E04Rik Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569891
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569891 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569891 Supplier Invitrogen™ Supplier No. PA569891
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 100%.

5730409E04Rik is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 5730409E04Rik
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene 5730409E04Rik
Gene Accession No. Q8BP99
Gene Alias 5730409E04Rik; 7530403E16Rik; AI849033; RIKEN cDNA 5730409E04Rik gene; UPF0500 protein C1orf216 homolog
Gene Symbols 5730409E04Rik
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse 5730409E04Rik (aa 178-227).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230757
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.