Learn More
Eppendorf™ 5804/5810 Series Four-Place Swinging Bucket Tube Adapters
Shop All Eppendorf North America Products
Description
Adapters are sold in packs of two. Order the proper number of adapters to fully equip your rotor.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| For Use With (Equipment) | A-4-44 swing bucket rotor for 5804/5804R/5810/5810R centrifuge |
| Bore Size (Metric) | 3.1 cm |