Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
58K Golgi Protein Antibody (FTCD/357), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314274R
This item is not returnable.
View return policy
Description
58K Golgi Protein Monoclonal antibody specifically detects 58K Golgi Protein in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
58K Golgi Protein | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot, Flow Cytometry, Immunofluorescence | |
FTCD/357 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
formimidoyltransferase cyclodeaminase, formimidoyltransferase-cyclodeaminase, formiminotransferase cyclodeaminase, Formiminotransferase-cyclodeaminase, human formiminotransferase cyclodeaminase, EC 4.3.1.410formiminotransferase-cyclodeaminase, LCHC1 | |
58K Golgi Protein Protein purified from rat liver. (Uniprot: O95954) | |
0.1 mL | |
Cellular Markers, Golgi Apparatus Markers, Lipid and Metabolism, Signal Transduction | |
10841 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction