Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5HT1D Receptor/SR-1D Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981897
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1897 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1897 Supplier Bioss Supplier No. BS12046R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

G-protein coupled receptor for serotonin; found in nociceptors throughout the body.

Specifications

Antigen 5HT1D Receptor/SR-1D
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HTR1D
Gene Accession No. P28221
Gene Alias 5 - Hydroxytryptamine (serotonin) receptor 1D; 5HT1D; 5-HT1D; 5-HT-1D; 5HT1D Receptor; 5-HT1d receptor; 5-HT-1D-alpha; 5-HT1DR; 5-hydroxytryptamine (serotonin) receptor 1D; 5-hydroxytryptamine (serotonin) receptor 1D beta; 5-hydroxytryptamine (serotonin) receptor 1D, G protein-coupled; 5-hydroxytryptamine receptor 1D; AI853647; Gpcr14; G-protein coupled receptor 14; HT1d receptor; HT1DA; Htr1d; HTR1DA; Htr1db; HTRL; RDC4; RP23-32L5.3; Serotonin 1D alpha receptor; Serotonin receptor 1D
Gene Symbols HTR1D
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human 5HT1D Receptor/SR-1D.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3352
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.