Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5HT3D Receptor Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12053R

Description
This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor is a ligand-gated ion channel, which when activated causes fast, depolarizing responses. It is a cation-specific, but otherwise relatively nonselective, ion channel.Specifications
5HT3D Receptor | |
Polyclonal | |
Unconjugated | |
HTR3D | |
5HT3D; 5-HT3-D; 5-hydroxytryptamine (serotonin) receptor 3 family member D; 5-hydroxytryptamine (serotonin) receptor 3D, ionotropic; 5-hydroxytryptamine receptor 3 subunit D; 5-hydroxytryptamine receptor 3D; HTR3D; serotonin 5-HT-3D receptor; Serotonin receptor 3D | |
Rabbit | |
Protein A | |
RUO | |
200909 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q70Z44 | |
HTR3D | |
KLH conjugated synthetic peptide derived from human 5HT3D Receptor. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction