Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5HT4 Receptor Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981905
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1905 Supplier Bioss Supplier No. BS12054R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is a member of the family of serotonin receptors, which are G protein coupled receptors that stimulate cAMP production in response to serotonin (5-hydroxytryptamine). The gene product is a glycosylated transmembrane protein that functions in both the peripheral and central nervous system to modulate the release of various neurotransmitters. Multiple transcript variants encoding proteins with distinct C-terminal sequences have been described, but the full-length nature of some transcript variants has not been determined.

Specifications

Antigen 5HT4 Receptor
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HTR4
Gene Accession No. Q13639
Gene Alias 5 hydroxytryptamine ( serotonin) receptor 4; 5 hydroxytryptamine (serotonin) receptor 4; 5-HT<4L>; 5HT4; 5-HT4; 5-HT-4; 5HT4 receptor; 5-HT4 receptor; 5-HT4 receptor (two splicing variants L and S); 5-HT4R; 5HTR4; 5-hydroxytryptamine (serotonin) receptor 4, G protein-coupled; 5-hydroxytryptamine receptor 4; cardiac 5-HT4 receptor; HTR4; Serotonin receptor 4
Gene Symbols HTR4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human 5HT4 Receptor.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3360
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.