Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 5J 4
GREENER_CHOICE

Catalog No. 549010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
549010 10 mg
549050 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 549010 Supplier Tocris Bioscience™ Supplier No. 5490/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

CRAC channel blocker; anti-inflammatory

5J 4 is a calcium release-activated calcium (CRAC) channel blocker. Blocks store-operated calcium entry (SOCE) in HeLa-O+S cells and Th17 cells in vitro. Ameliorates symptoms and reduces CNS infiltration by mononuclear cells in a mouse model of experimental autoimmune encephalomyelitis (EAE) in vivo.

Specifications

Chemical Name or Material N-[[(6-Hydroxy-1-naphthalenyl)amino]thioxomethyl]-2-furancarboxamide
CAS 827001-82-1
Quantity 10 mg
Target Other Calcium Channel Blockers
Molecular Formula C16H12N2O3S
Formula Weight 312.34
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.