Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5T4 Antibody (13C3), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | 5T4 |
---|---|
Clone | 13C3 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
5T4 Monoclonal antibody specifically detects 5T4 in Human, Macaque samples. It is validated for ELISASpecifications
5T4 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human, Macaque | |
5T4 oncofetal trophoblast glycoprotein, 5T4 oncotrophoblast glycoprotein, 5T4AG, 5T4-AG, 5T4M6P1,5T4 oncofetal antigen, trophoblast glycoprotein | |
Recombinant Human 5T4 protein [UniProt Q13641] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
13C3 | |
ELISA | |
Unconjugated | |
Human | |
Cancer, Cell Biology, Immunology, Stem Cells | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
7162 | |
IgG1 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title