Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5T4 Antibody (1D4), Novus Biologicals™
SDP

Catalog No. p-200022767 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298426 50 μL
NB298427 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298426 Supplier Novus Biologicals Supplier No. NBP32627550UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

5T4 Monoclonal antibody specifically detects 5T4 in Human samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen 5T4
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 1D4
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias 5T4 oncofetal trophoblast glycoprotein, 5T4 oncotrophoblast glycoprotein, 5T4AG, 5T4-AG, 5T4M6P1,5T4 oncofetal antigen, trophoblast glycoprotein
Host Species Rabbit
Immunogen A synthesized peptide derived from Human 5T4 [UniProt Q13641]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 7162
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.