Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5T4 Antibody (524744), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB49751D
Description
5T4 Monoclonal antibody specifically detects 5T4 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
5T4 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
524744 | |
Flow Cytometry, CyTOF-ready | |
5T4 oncofetal trophoblast glycoprotein, 5T4 oncotrophoblast glycoprotein, 5T4AG, 5T4-AG, 5T4M6P1,5T4 oncofetal antigen, trophoblast glycoprotein | |
Mouse myeloma cell line NS0-derived recombinant human 5T4, Ser31-Ser355, Accession # Q13641 | |
0.1 mL | |
Cancer, Cell Biology, Immunology, Stem Cells | |
7162 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction