Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 5T4 Monoclonal Antibody (524744)
GREENER_CHOICE

Catalog No. PIMA524229
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524229 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524229 Supplier Invitrogen™ Supplier No. MA524229
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In direct ELISAs, no cross-reactivity with recombinant mouse 5T4 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

This gene encodes a leucine-rich transmembrane glycoprotein that may be involved in cell adhesion. The encoded protein is an oncofetal antigen that is specific to trophoblast cells. In adults this protein is highly expressed in many tumor cells and is associated with poor clinical outcome in numerous cancers. Alternate splicing in the 5' UTR results in multiple transcript variants that encode the same protein. [provided by RefSeq, Oct 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen 5T4
Applications Flow Cytometry
Classification Monoclonal
Clone 524744
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene TPBG
Gene Accession No. Q13641
Gene Alias 5T4; 5T4 oncofetal antigen; 5T4 oncofetal trophoblast glycoprotein; 5T4 oncotrophoblast glycoprotein; 5T4AG; 5T4-AG; AW495680; M6P1; TPBG; Trophoblast glycoprotein; WAIF1; Wnt-activated inhibitory factor 1
Gene Symbols TPBG
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human 5T4 Ser31-Ser355.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7162
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.