Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 5T4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118921
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118921 200 μL
PIPA5122307 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5118921 Supplier Invitrogen™ Supplier No. PA5118921
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

5T4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes a leucine-rich transmembrane glycoprotein that may be involved in cell adhesion. The encoded protein is an oncofetal antigen that is specific to trophoblast cells. In adults this protein is highly expressed in many tumor cells and is associated with poor clinical outcome in numerous cancers. Alternate splicing in the 5' UTR results in multiple transcript variants that encode the same protein. [provided by RefSeq, Oct 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen 5T4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene TPBG
Gene Accession No. Q13641, Q9Z0L0
Gene Alias 5T4; 5T4 oncofetal antigen; 5T4 oncofetal trophoblast glycoprotein; 5T4 oncotrophoblast glycoprotein; 5T4AG; 5T4-AG; AW495680; M6P1; TPBG; Trophoblast glycoprotein; WAIF1; Wnt-activated inhibitory factor 1
Gene Symbols TPBG
Host Species Rabbit
Immunogen Recombinant protein 5T4. The antigen corresponds to amino acid range 32-351 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21983, 7162
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.