Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ 5T4 Polyclonal Antibody

Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA547679
Description
In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse 5T4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a leucine-rich transmembrane glycoprotein that may be involved in cell adhesion. The encoded protein is an oncofetal antigen that is specific to trophoblast cells. In adults this protein is highly expressed in many tumor cells and is associated with poor clinical outcome in numerous cancers. Alternate splicing in the 5' UTR results in multiple transcript variants that encode the same protein. [provided by RefSeq, Oct 2009].
Specifications
| 5T4 | |
| Polyclonal | |
| Unconjugated | |
| TPBG | |
| 5T4; 5T4 oncofetal antigen; 5T4 oncofetal trophoblast glycoprotein; 5T4 oncotrophoblast glycoprotein; 5T4AG; 5T4-AG; AW495680; M6P1; TPBG; Trophoblast glycoprotein; WAIF1; Wnt-activated inhibitory factor 1 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 7162 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q13641 | |
| TPBG | |
| Mouse myeloma cell line NS0-derived recombinant human 5T4 Ser31-Ser355. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction