Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 5T4 Recombinant Rabbit Monoclonal Antibody (SU34-05)
GREENER_CHOICE

Catalog No. PIMA532120
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532120 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532120 Supplier Invitrogen™ Supplier No. MA532120
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

This gene encodes a leucine-rich transmembrane glycoprotein that may be involved in cell adhesion. The encoded protein is an oncofetal antigen that is specific to trophoblast cells. In adults this protein is highly expressed in many tumor cells and is associated with poor clinical outcome in numerous cancers. Alternate splicing in the 5' UTR results in multiple transcript variants that encode the same protein. [provided by RefSeq, Oct 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen 5T4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone SU34-05
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% Glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene TPBG
Gene Accession No. Q13641, Q5PQV5, Q9Z0L0
Gene Alias 5T4; 5T4 oncofetal antigen; 5T4 oncofetal trophoblast glycoprotein; 5T4 oncotrophoblast glycoprotein; 5T4AG; 5T4-AG; AW495680; M6P1; TPBG; Trophoblast glycoprotein; WAIF1; Wnt-activated inhibitory factor 1
Gene Symbols TPBG
Host Species Rabbit
Immunogen Synthetic peptide within Human 5T4 aa 381-420.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21983, 7162, 83684
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.