Learn More
Promega 5X PCR Buffer E, 1 each
Catalog No. PRD2341 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 ea.
Catalog No. Quantity
PRD2341 1 ea.
1 options
This item is not returnable. View return policy
Catalog No. PRD2341 Supplier Promega Supplier No. D2341
This item is not returnable. View return policy
5X PCR Buffers are preformulated, high-quality buffers, 25mM MgCl2 and GoTaq MDx Hot Start Polymerase.
  • Individual, Preformulated PCR Optimization Kit Components
  • 5X PCR buffers included in PCR Optimization Kit, available individually
  • Manufactured under cGMP for consistent, lot−to−lot PCR performance
  • Stand−alone buffer supplied with MgCl2 and GoTaq(R) MDx Hot Start Polymerase
Specifications
Quantity 1 ea.