Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
(5Z)-7-Oxozeaenol is a resorcyclic lactone of fungal origin that acts as a potent and selective transforming growth factor-β-activated kinase 1 (TAK1) mitogen-activated protein kinase kinase kinase (MAPKKK) inhibitor (IC50 = 8 nM). Displays > 33-fold and > 62-fold selectivity over MEKK1 and MEKK4 respectively. Inhibits IL-1-induced activation of NF-κB (IC50 = 83 nM) and JNK/p38. Inhibits production of inflammatory mediators, and sensitizes cells to TRAIL- and TNF-α-induced apoptosis in vitro.
Specifications
Specifications
| Quantity | 1 mg |
| Product Type | (5Z)-7-Oxozeaenol |
| Purity | >99% |
| Molecular Weight (g/mol) | 362.37 |
| Inhibitors | (5Z)-7-Oxozeaenol |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?

