Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6(5H)-Phenanthridinone, 96%

Catalog No. p-7028064
Click to view available options
Quantity:
1 g
5 g

CAS: 1015-89-0 | C13H9NO | 195.221 g/mol

6(5H)-Phenanthridinone is used as a probe into the role of PARP in cellular response to irradiation. Treatment of cells with 6(5H)-Phenanthridinone and various DNA-damaging agents showed a resistance to apoptosis. It acts as a reactant in the synthesis of 5,6-dihydrophenanthridine sulfonamides, oxidative coupling with diphenylacetylene, direct copper acetate-catalyzed N-cyclopropylation of cyclic amides. It is used as HIV-1 integrase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6(5H)-Phenanthridinone is used as a probe into the role of PARP in cellular response to irradiation. Treatment of cells with 6(5H)-Phenanthridinone and various DNA-damaging agents showed a resistance to apoptosis. It acts as a reactant in the synthesis of 5,6-dihydrophenanthridine sulfonamides, oxidative coupling with diphenylacetylene, direct copper acetate-catalyzed N-cyclopropylation of cyclic amides. It is used as HIV-1 integrase inhibitor.

Solubility
Soluble in DMSO (5 mg/ml). Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1015-89-0
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.221
MDL Number MFCD00004988
InChI Key RZFVLEJOHSLEFR-UHFFFAOYSA-N
Synonym 6 5h-phenanthridinone, phenanthridin-6 5h-one, 6-phenanthridinol, phenanthridone, 6-5h-phenanthridinone, 6-phenanthridone, 6-phenanthridinone, 6 5h-phenanthridone, phenanthridin-6-ol, phenantridone
PubChem CID 1853
ChEBI CHEBI:75292
IUPAC Name 5H-phenanthridin-6-one
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3NC2=O

Specifications

Melting Point 288°C to 294°C
Quantity 1 g
Beilstein 140641
Solubility Information Soluble in DMSO (5mg/ml). Insoluble in water.
Formula Weight 195.22
Percent Purity 96%
Chemical Name or Material 6(5H)-Phenanthridinone
EINECSNumber 213-804-3
RTECSNumber SG0370000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.