Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
6,6,7,7,8,8,8-Heptafluoro-5,5-bis(trifluoromethyl)-1,3-octadiene 98.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Color | Yellow |
Quantity | 25 g |
UN Number | 1993 |
Formula Weight | 372.13 |
Percent Purity | ≥98.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | 6,6,7,7,8,8,8-Heptafluoro-5,5-bis(trifluoromethyl)-1,3-octadiene |
Safety and Handling
TSCA | No |
Recommended Storage | Refrigerator |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction