Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,6,7,7,8,8,8-Heptafluoro-5,5-bis(trifluoromethyl)-1,3-octadiene 98.0+%, TCI America™
SDP

Catalog No. H15155G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1429188-77-1
Molecular Formula C10H5F13
Molecular Weight (g/mol) 372.129
MDL Number MFCD28014057
InChI Key PUTFFSRJABOVAD-ONEGZZNKSA-N
PubChem CID 122360969
IUPAC Name (3E)-6,6,7,7,8,8,8-heptafluoro-5,5-bis(trifluoromethyl)octa-1,3-diene
SMILES C=CC=CC(C(C(C(F)(F)F)(F)F)(F)F)(C(F)(F)F)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
UN Number 1993
Formula Weight 372.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 6,6,7,7,8,8,8-Heptafluoro-5,5-bis(trifluoromethyl)-1,3-octadiene
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.