Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,6'-Dibromo-9,9'-diphenyl-3,3'-bicarbazole 95.0+%, TCI America™
SDP

Catalog No. D49801G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49801G 1 g
D4980200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49801G Supplier TCI America Supplier No. D49801G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 354135-75-4
Molecular Formula C36H22Br2N2
Molecular Weight (g/mol) 642.394
InChI Key VTIPHPNQFUCDAW-UHFFFAOYSA-N
PubChem CID 131852839
IUPAC Name 3-bromo-6-(6-bromo-9-phenylcarbazol-3-yl)-9-phenylcarbazole
SMILES C1=CC=C(C=C1)N2C3=C(C=C(C=C3)C4=CC5=C(C=C4)N(C6=C5C=C(C=C6)Br)C7=CC=CC=C7)C8=C2C=CC(=C8)Br

Specifications

Quantity 1 g
Formula Weight 642.39
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 6,6′-Dibromo-9,9′-diphenyl-3,3′-bicarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.