Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
6,6'-Dimethyl-2,2'-bipyridyl 98.0+%, TCI America™

Click to view available options
Quantity:
100 mg
1 g
5 g
Specifications
Specifications
Melting Point | 88°C |
Color | White-Yellow |
Boiling Point | 286°C |
Quantity | 100 mg |
Synonym | 6,6′C-Bi-2-picoline, 6,6′C-Di-2-picolyl |
Formula Weight | 184.24 |
Percent Purity | ≥98.0% (GC,T) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 6,6′-Dimethyl-2,2′-bipyridyl |
Safety and Handling
RTECSNumber | DW1767000 |
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction