Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,7,8,9-Tetrahydro-5H-benzocyclohepten-5-one 97.0+%, TCI America™
SDP

Catalog No. T134725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T134725G 25 g
T13475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T134725G Supplier TCI America Supplier No. T134725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 826-73-3
Molecular Formula C11H12O
Molecular Weight (g/mol) 160.22
MDL Number MFCD00004144
InChI Key KWHUHTFXMNQHAA-UHFFFAOYSA-N
Synonym 1-Benzosuberone
PubChem CID 70003
IUPAC Name 6,7,8,9-tetrahydro-5H-benzo[7]annulen-5-one
SMILES O=C1CCCCC2=CC=CC=C12

Specifications

Color Yellow
Boiling Point 270°C
Quantity 25 g
Formula Weight 160.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 6,7,8,9-Tetrahydro-5H-benzocyclohepten-5-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.