Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,8-Dibromoimidazo[1,2-a]pyrazine, 95%, Thermo Scientific Chemicals

Catalog No. p-7039755
Click to view available options
Quantity:
5 g

It is an Intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an Intermediate in organic syntheses.

Solubility
Slightly soluble in water.

Notes
Recommended storage temperature: 2 - 4°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63744-22-9
Molecular Formula C6H3Br2N3
Molecular Weight (g/mol) 276.92
MDL Number MFCD08460056
InChI Key UQCZZGIPIMJBCL-UHFFFAOYSA-N
Synonym 6,8-dibromoimidazo 1,2-a pyrazine, imidazo 1,2-a pyrazine, 6,8-dibromo, 6,8-dibromo-imidazo 1,2-a pyrazine, 6,8-dibromoimidazol 1,2-a pyrazine, pubchem14656, acmc-1b8pv, ksc351o9d, buttpark 180\02-82, 6,8 dibromoimidazo 1,2-a pyrazine, 6,8-dibomoimidazo 1,2-a pyrazine
PubChem CID 15025843
IUPAC Name 6,8-dibromoimidazo[1,2-a]pyrazine
SMILES BrC1=CN2C=CN=C2C(Br)=N1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 276.92
Percent Purity 95%
Chemical Name or Material 6,8-Dibromoimidazo[1,2-a]pyrazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.