Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
6,8-Dichloro-3-formylchromone, 97%, Thermo Scientific™
Click to view available options
:
5g; Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Specifications
CAS | 64481-10-3 |
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (HPLC) |
Packaging | Glass bottle |
Molecular Formula | C10H4Cl2O3 |
Quantity | 5g |
MDL Number | MFCD00051510 |
Synonym | 6,8-dichloro-3-formylchromone, 6,8-dichloro-4-oxo-4h-chromene-3-carbaldehyde, 6,8-dichlorochromone-3-carboxaldehyde, chembl85493, 6,8-dichloro-4-oxo-4h-1-benzopyran-3-carboxaldehyde, 4h-1-benzopyran-3-carboxaldehyde, 6,8-dichloro-4-oxo, pubchem8676, 6,8-dichlorochromon-3-aldehyde, 6,8-dichloro-4-oxo-chromene-3-carbaldehyde, 3-formyl-6,8-dichloro-4h-1-benzopyran-4-one |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction