Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-1-hexanol, 97%, Thermo Scientific Chemicals

Catalog No. AAL1415703
Click to view available options
Quantity:
1 g
5 g

6-Amino-1-hexanol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Amino-1-hexanol is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4048-33-3
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.192
MDL Number MFCD00008241
InChI Key SUTWPJHCRAITLU-UHFFFAOYSA-N
Synonym 6-amino-1-hexanol, 1-hexanol, 6-amino, 6-aminohexanol, 1-amino-6-hexanol, hexanol, 6-amino, 6-hydroxyhexylamine, aminohexyl alcohol, amidohexylalkohol, amidohexylalkohol german, 6-amino-hexan-1-ol
PubChem CID 19960
IUPAC Name 6-aminohexan-1-ol
SMILES C(CCCO)CCN

Specifications

Melting Point 55°C to 58°C
Boiling Point 136°C to 139°C (30 mmHg)
Flash Point 133°C (271°F)
Quantity 1 g
Beilstein 1732524
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 117.19
Percent Purity 97%
Chemical Name or Material 6-Amino-1-hexanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-748-1
RTECSNumber MO8840000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.