Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-2-methylquinoline, 97%

Catalog No. AAA1396703
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1396703 1 g
AAA13967MD 250 mg
AAA1396706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1396703 Supplier Thermo Scientific Chemicals Supplier No. A1396703
Add to Cart

CAS: 65079-19-8 | C10H10N2 | 158.204 g/mol

6-Amino-2-methylquinoline is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Amino-2-methylquinoline is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65079-19-8
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
MDL Number MFCD00052600
InChI Key TYJFYUVDUUACKX-UHFFFAOYSA-N
PubChem CID 103148
IUPAC Name 2-methylquinolin-6-amine
SMILES CC1=NC2=C(C=C1)C=C(C=C2)N

Specifications

Melting Point 189°C to 191°C
Quantity 1 g
Beilstein 114795
Synonym 6-amino-2-methylquinoline, 2-methyl-6-quinolinamine, 6-quinolinamine, 2-methyl, 6-aminoquinaldine, 2-methyl-6-aminoquinoline, 6-quinolinamine,2-methyl, 2-methyl-quinolin-6-ylamine, 2-methyl-6-quinolylamine, mqn, 2-methylquinoline-6-amine
Solubility Information Soluble in water.
Formula Weight 158.2
Percent Purity 97%
Chemical Name or Material 6-Amino-2-methylquinoline
Hazard Category H302-H315-H319-H335-H500
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.