Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-5-bromoquinoline, 97%

Catalog No. AAH63150MD
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 50358-42-4 | C9H7BrN2 | 223.073 g/mol

6-Amino-5-bromoquinoline, has a wide range of biological and pharmacological activities. It shows both electrophilic and nucleophilic substitution reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Amino-5-bromoquinoline, has a wide range of biological and pharmacological activities. It shows both electrophilic and nucleophilic substitution reactions.

Solubility
Slightly soluble in water. Soluble in alcohol, ether, and carbon disulfide.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50358-42-4
Molecular Formula C9H7BrN2
Molecular Weight (g/mol) 223.073
MDL Number MFCD00234503
InChI Key MODLGTLYXJGDCH-UHFFFAOYSA-N
PubChem CID 12741232
IUPAC Name 5-bromoquinolin-6-amine
SMILES C1=CC2=C(C=CC(=C2Br)N)N=C1

Specifications

Color Yellow to Brown
Quantity 250 mg
Synonym 6-amino-5-bromoquinoline, 6-quinolinamine, 5-bromo, 5-bromo-6-quinolinamine, 6-quinolinamine,5-bromo, 5-bromo-quinolin-6-ylamine, 6-quinolinamine, 5-bromo-9ci
Solubility Information Slightly soluble in water. Soluble in alcohol,ether,and carbon disulfide.
Formula Weight 223.07
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Amino-5-bromoquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.