Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(Aminomethyl)quinoline, 95%, Thermo Scientific Chemicals

Catalog No. AAH6654803
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6654803 1 g
AAH66548MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6654803 Supplier Thermo Scientific Chemicals Supplier No. H6654803
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99071-54-2
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.20
MDL Number MFCD02853688
InChI Key RZIPENSSTUBRAA-UHFFFAOYSA-N
Synonym 6-aminomethylquinoline, 6-quinolinemethanamine, 6-aminomethyl quinoline, quinolin-6-yl methanamine, quinolin-6-ylmethyl amine, quinoline-6-ylmethanamine, 1-quinolin-6-yl methanamine, 6-quinolylmethylamine, c-quinolin-6-yl-methylamine, 6-quinolylmethanamine
PubChem CID 1514384
IUPAC Name 1-(quinolin-6-yl)methanamine
SMILES NCC1=CC2=CC=CN=C2C=C1

Specifications

Quantity 1 g
Beilstein 115231
Formula Weight 158.2
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 6-(Aminomethyl)quinoline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.